Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001725.2 | NP_001716.2 | 13 | 1377 | ||
GenBank mRNA | AF322588.1 | - | 13 | 1303 | ||
AK296568.1 | - | 13 | 1222 | |||
AK315328.1 | - | 13 | 1344 | |||
AX713278.1 | - | 13 | 1345 | |||
AX746123.1 | - | 13 | 1345 | |||
AX752831.1 | - | 13 | 1345 | |||
BC040955.1 | - | 13 | 1352 | 5218655 | ||
DQ414688.1 | - | 13 | 1315 | |||
J04739.1 | - | 13 | 1345 |