Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042550.1 | NP_001036015.1 | 20 | 3092 | ||
NM_001042551.1 | NP_001036016.1 | 20 | 3030 | |||
NM_001265602.1 | NP_001252531.1 | 20 | 3046 | |||
NM_006444.2 | NP_006435.2 | 20 | 3021 | |||
XM_006716933.3 | XP_006716996.1 | 20 | 2943 | |||
XM_011518148.2 | XP_011516450.1 | 20 | 2906 | |||
XM_011518149.2 | XP_011516451.1 | 20 | 2948 | |||
XM_011518150.2 | XP_011516452.1 | 19 | 2810 | |||
XM_011518151.2 | XP_011516453.1 | 19 | 2788 | |||
XM_011518152.2 | XP_011516454.1 | 20 | 2972 | |||
XM_011518153.1 | XP_011516455.1 | 20 | 2972 | |||
XM_017014206.1 | XP_016869695.1 | 20 | 3092 | |||
XM_017014207.1 | XP_016869696.1 | 20 | 2994 | |||
XM_017014208.1 | XP_016869697.1 | 20 | 3034 | |||
XM_017014209.1 | XP_016869698.1 | 19 | 2921 | |||
XM_017014210.1 | XP_016869699.1 | 19 | 2811 | |||
XM_017014211.1 | XP_016869700.1 | 19 | 2900 | |||
XM_017014212.1 | XP_016869701.1 | 19 | 2767 | |||
GenBank mRNA | AF113673.1 | - | 1 | 26 | ||
AK000955.1 | - | 9 | 1263 | |||
AK001485.1 | - | 20 | 3078 | |||
AK292599.1 | - | 20 | 2969 | |||
AL833191.1 | - | 20 | 3031 | |||
BC130385.1 | - | 20 | 2876 | 40146416 | ||
BC144163.1 | - | 20 | 2892 | 9052683 |