Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321441.1 | NP_001308370.1 | 2, 3 | 301 | ||
NM_012111.2 | NP_036243.1 | 2, 3 | 435 | |||
GenBank mRNA | AF161440.1 | - | 1, 2 | 79 | ||
AF164791.1 | - | 2, 3 | 341 | |||
AJ243310.1 | - | 2, 3 | 406 | |||
AK300766.1 | - | 2, 3 | 433 | |||
AK313824.1 | - | 2, 3 | 369 | |||
BC000321.2 | - | 2, 3 | 383 | 2821759 | ||
BC007398.2 | - | 1, 2 | 47 | |||
BG432594.1 | - | 2, 3 | 436 | |||
BQ073652.1 | - | 2, 3 | 225 | |||
CX786861.1 | - | 2, 3 | 312 |