Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAICS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079524.1 | NP_001072992.1 | 7 | 1110 | ||
NM_001079525.1 | NP_001072993.1 | 8 | 1131 | |||
NM_006452.3 | NP_006443.1 | 8 | 1131 | |||
XM_011534366.1 | XP_011532668.1 | 7 | 951 | |||
GenBank mRNA | AK294821.1 | - | 8 | 1175 | ||
AK308527.1 | - | 7 | 984 | |||
BC010273.1 | - | 8 | 1105 | 2900848 | ||
BC019255.1 | - | 7 | 1061 | 3533684 | ||
BM807012.1 | - | 4 | 498 | |||
BT006988.1 | - | 7 | 900 | |||
CR749824.1 | - | 8 | 1096 | |||
X53793.1 | - | 7 | 924 |