Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006453.2 | NP_006444.2 | 7, 8 | 746 | ||
GenBank mRNA | AK095076.1 | - | 2, 3 | 221 | ||
BC010231.1 | - | 7, 8 | 746 | 3455740 | ||
BC014007.2 | - | 7, 8 | 718 | 4040312 | ||
BC035409.1 | - | 7, 8 | 702 | 5165833 | ||
U02609.1 | - | 7, 8 | 694 |
Set Membership: |
G-protein coupled receptor |