Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248006.1 | NP_001234935.1 | 8 | 2026 | ||
NM_001248007.1 | NP_001234936.1 | 7 | 1689 | |||
NM_006458.3 | NP_006449.2 | 9 | 2027 | |||
NM_033278.3 | NP_150594.2 | 8 | 1862 | |||
XM_011519844.1 | XP_011518146.1 | 9 | 2126 | |||
XM_011519845.2 | XP_011518147.1 | 8 | 1807 | |||
XM_017017098.1 | XP_016872587.1 | 8 | 1838 | |||
XM_017017099.1 | XP_016872588.1 | 8 | 1730 | |||
GenBank mRNA | AF045239.1 | - | 7 | 1631 | ||
AF220020.1 | - | 7 | 1631 | |||
AF220021.1 | - | 8 | 1598 | |||
AF220022.1 | - | 7 | 1394 | |||
AK122682.1 | - | 1 | 629 | |||
AK126685.1 | - | 6 | 1255 | |||
AK127278.1 | - | 1 | 1905 | |||
AK291396.1 | - | 8 | 2026 | |||
AK298843.1 | - | 7 | 1565 | |||
AK299586.1 | - | 8 | 1819 | |||
BC096827.1 | - | 8 | 1862 | 6108991 |