Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248006.1 | NP_001234935.1 | 7 | 1917 | ||
NM_001248007.1 | NP_001234936.1 | 6 | 1580 | |||
NM_006458.3 | NP_006449.2 | 8 | 1918 | |||
NM_033278.3 | NP_150594.2 | 7 | 1753 | |||
XM_011519844.1 | XP_011518146.1 | 8 | 2017 | |||
XM_011519845.2 | XP_011518147.1 | 7 | 1698 | |||
XM_017017098.1 | XP_016872587.1 | 7 | 1729 | |||
XM_017017099.1 | XP_016872588.1 | 7 | 1621 | |||
GenBank mRNA | AF045239.1 | - | 6 | 1522 | ||
AF220020.1 | - | 6 | 1522 | |||
AF220021.1 | - | 7 | 1489 | |||
AF220022.1 | - | 6 | 1285 | |||
AK126685.1 | - | 5 | 1146 | |||
AK291396.1 | - | 7 | 1917 | |||
AK298843.1 | - | 6 | 1456 | |||
AK299586.1 | - | 7 | 1710 | |||
BC096827.1 | - | 7 | 1753 | 6108991 |