Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006461.3 | NP_006452.3 | 14, 15 | 2633 | ||
GenBank mRNA | AF063308.1 | - | 14, 15 | 2610 | ||
AF345347.2 | - | 14, 15 | 2874 | |||
AF399910.1 | - | 14, 15 | 2612 | |||
AK025725.1 | - | 14, 15 | 2604 | |||
AK292398.1 | - | 14, 15 | 2624 | |||
AL122116.1 | - | 3, 4 | 320 | |||
AL137585.1 | - | 12, 13 | 2044 | |||
BC000322.2 | - | 14, 15 | 2593 | 2821784 |