Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206840.1 | NP_001193769.1 | 2 | 1088 | ||
NM_001206841.1 | NP_001193770.1 | 2 | 1088 | |||
NM_006464.3 | NP_006455.2 | 2 | 1088 | |||
GenBank mRNA | AF027515.1 | - | 2 | 816 | ||
AF027516.1 | - | 2 | 816 | |||
AK222829.1 | - | 2 | 780 | |||
AK223063.1 | - | 2 | 779 | |||
AK312479.1 | - | 2 | 781 | |||
BC008461.1 | - | 2 | 781 | 4272702 | ||
BC028219.1 | - | 2 | 777 | 5217823 | ||
U62390.1 | - | 2 | 816 | |||
X94333.1 | - | 2 | 772 |