Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BRDT SIRNA Assays ›
Gene Symbol:
BRDT
Gene Name:
bromodomain testis associated
Gene Aliases:
BRD6, CT9
Chromosome Location:
Chr.1: 91949371 - 92014428 on Build GRCh38
UniGene ID:
Hs.482520
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2088
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BRDT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242805.2
NP_001229734.2
8, 9
1524
NM_001242806.2
NP_001229735.2
7, 8
1467
NM_001242807.2
NP_001229736.2
6, 7
1123
NM_001242808.2
NP_001229737.2
6, 7
1317
NM_001242810.2
NP_001229739.2
7, 8
1811
NM_001726.4
NP_001717.3
7, 8
1250
NM_207189.3
NP_997072.2
7, 8
1455
XM_006710853.3
XP_006710916.1
8, 9
2040
XM_006710854.3
XP_006710917.1
9, 10
1885
XM_006710855.3
XP_006710918.1
9, 10
1890
XM_006710856.3
XP_006710919.1
9, 10
1864
XM_006710857.3
XP_006710920.1
9, 10
1854
XM_011542032.2
XP_011540334.1
9, 10
1374
XM_011542033.2
XP_011540335.1
9, 10
1348
XM_011542034.2
XP_011540336.1
9, 10
1364
XM_011542035.2
XP_011540337.1
9, 10
1338
XM_011542036.2
XP_011540338.1
7, 8
1253
GenBank mRNA
AF019085.1
-
7, 8
1213
AK302758.1
-
6, 7
1059
AK303008.1
-
7, 8
1215
AK316442.1
-
6, 7
1122
AY338951.1
-
8, 9
1524
BC005281.1
-
7, 8
1181
BC017582.1
-
7, 8
1256
BC047900.1
-
7, 8
1220
BC062700.1
-
7, 8
1215
BX648135.1
-
7, 8
1546
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
histone displacement
chromatin remodeling
transcription, DNA-templated
mRNA processing
male meiosis
male meiosis I
spermatogenesis
RNA splicing
covalent chromatin modification
cell differentiation
regulation of RNA splicing
positive regulation of transcription involved in meiotic cell cycle
transcription coactivator activity
histone binding
lysine-acetylated histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0