Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313972.1 | NP_001300901.1 | 6 | 1022 | ||
NM_006472.5 | NP_006463.3 | 7 | 1391 | |||
XM_017000085.1 | XP_016855574.1 | 7 | 1340 | |||
GenBank mRNA | AK226090.1 | - | 7 | 1303 | ||
AK301148.1 | - | 7 | 913 | |||
AK301203.1 | - | 3 | 511 | |||
AK304670.1 | - | 6 | 1022 | |||
AY594328.2 | - | 7 | 1026 | |||
BC024305.1 | - | 7 | 920 | |||
BC093702.1 | - | 7 | 1100 | 7939547 | ||
BC093704.1 | - | 7 | 1100 | 7939549 | ||
BX537824.1 | - | 4 | 773 | |||
S73591.1 | - | 7 | 1247 |