Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006473.3 | NP_006464.1 | 11 | 1883 | ||
XM_005273714.1 | XP_005273771.1 | 11 | 1748 | |||
XM_017017100.1 | XP_016872589.1 | 4 | 1182 | |||
GenBank mRNA | AF069735.1 | - | 10 | 1682 | ||
AK289499.1 | - | 11 | 1793 | |||
AK314330.1 | - | 11 | 1769 | |||
BC008785.2 | - | 11 | 1783 |