Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135934.1 | NP_001129406.1 | 2 | 299 | ||
NM_001135935.1 | NP_001129407.1 | 2 | 299 | |||
NM_001135936.1 | NP_001129408.1 | 2 | 299 | |||
NM_001286665.1 | NP_001273594.1 | 2 | 299 | |||
NM_001286666.1 | NP_001273595.1 | 2 | 299 | |||
NM_001286667.1 | NP_001273596.1 | 2 | 299 | |||
NM_006475.2 | NP_006466.2 | 2 | 299 | |||
XM_005266231.2 | XP_005266288.1 | 2 | 294 | |||
XM_005266232.2 | XP_005266289.1 | 2 | 294 | |||
XM_017020355.1 | XP_016875844.1 | 2 | 294 | |||
XM_017020356.1 | XP_016875845.1 | 2 | 294 | |||
GenBank mRNA | AK300261.1 | - | 2 | 413 | ||
AY140646.1 | - | 2 | 192 | |||
AY918092.1 | - | 2 | 192 | |||
BC106709.1 | - | 2 | 192 | 40007999 | ||
BC106710.1 | - | 2 | 192 | 40008000 | ||
BY795054.2 | - | 2 | 206 | |||
D13665.1 | - | 2 | 208 | |||
D13666.1 | - | 2 | 192 | |||
DC403736.1 | - | 2 | 299 | |||
EU262883.1 | - | 2 | 181 | |||
EU262884.1 | - | 2 | 181 | |||
EU262885.1 | - | 2 | 181 | |||
EU262886.1 | - | 2 | 181 | |||
GU354210.1 | - | 2 | 192 |