Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAS2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278730.1 | NP_001265659.1 | 4 | 968 | ||
NM_006478.4 | NP_006469.2 | 4 | 996 | |||
NM_152236.2 | NP_689422.1 | 4 | 1001 | |||
XM_011529824.1 | XP_011528126.1 | 5 | 1120 | |||
XM_011529825.1 | XP_011528127.1 | 4 | 929 | |||
XM_017028533.1 | XP_016884022.1 | 4 | 1646 | |||
GenBank mRNA | AK093386.1 | - | 4 | 938 | ||
AK122829.1 | - | 3 | 1838 | |||
AK223602.1 | - | 4 | 940 | |||
AK225484.1 | - | 4 | 940 | |||
AK301715.1 | - | 3 | 423 | |||
BC001782.1 | - | 4 | 936 | 3355168 | ||
BC007624.1 | - | 4 | 936 | 3357387 | ||
BC011047.1 | - | 4 | 939 | 4178571 | ||
Y07846.1 | - | 4 | 922 |