Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD51AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130862.1 | NP_001124334.1 | 6 | 582 | ||
NM_006479.4 | NP_006470.1 | 5 | 531 | |||
GenBank mRNA | AF006259.1 | - | 5 | 426 | ||
AK096930.1 | - | 6 | 459 | |||
AK290428.1 | - | 6 | 476 | |||
AK291948.1 | - | 5 | 425 | |||
AK300773.1 | - | 4 | 315 | |||
AK316367.1 | - | 4 | 329 | |||
AL832243.1 | - | 6 | 460 | |||
BC006992.1 | - | 5 | 425 | 3997352 | ||
BC016330.1 | - | 6 | 474 | 4107592 | ||
DC397655.1 | - | 5 | 531 |