Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006480.4 | NP_006471.2 | 4 | 459 | ||
XM_005265794.4 | XP_005265851.1 | 4 | 626 | |||
XM_005265795.4 | XP_005265852.1 | 4 | 631 | |||
GenBank mRNA | BC014094.2 | - | 4 | 429 | 4547415 | |
BI752372.1 | - | 4 | 465 |