Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271998.1 | NP_001258927.1 | 6 | 2163 | ||
NM_004578.3 | NP_004569.2 | 8 | 2397 | |||
NR_073545.1 | 8 | 2359 | ||||
GenBank mRNA | AY585832.1 | - | 8 | 2327 |
Gene Symbol: SPHAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006542.3 | NP_006533.1 | 1 | 874 | ||
GenBank mRNA | BC070287.1 | - | 1 | 399 | 30320725 | |
BC107681.1 | - | 1 | 874 |