Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMVK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323011.1 | NP_001309940.1 | 5 | 485 | ||
NM_001323012.1 | NP_001309941.1 | 5 | 492 | |||
NM_006556.3 | NP_006547.1 | 5 | 835 | |||
GenBank mRNA | AK130234.1 | - | 6 | 723 | ||
AK312803.1 | - | 5 | 794 | |||
BC006089.1 | - | 5 | 818 | 3604167 | ||
BC007694.2 | - | 5 | 748 | 3954909 | ||
BT019976.1 | - | 5 | 512 | |||
CR541998.1 | - | 5 | 512 | |||
L77213.1 | - | 5 | 547 |