Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF2 SIRNA Assays ›
Gene Symbol:
CELF2
Gene Name:
CUGBP, Elav-like family member 2
Gene Aliases:
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location:
Chr.10: 10462515 - 11336675 on Build GRCh38
UniGene ID:
Hs.309288
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20958
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025076.2
NP_001020247.1
11
1416
NM_001025077.2
NP_001020248.1
11
1255
NM_001083591.1
NP_001077060.1
11
1410
NM_001326317.1
NP_001313246.1
13
1257
NM_001326318.1
NP_001313247.1
13
1275
NM_001326319.1
NP_001313248.1
15
1367
NM_001326320.1
NP_001313249.1
14
1369
NM_001326321.1
NP_001313250.1
14
1299
NM_001326323.1
NP_001313252.1
15
1411
NM_001326324.1
NP_001313253.1
12
1252
NM_001326325.1
NP_001313254.1
13
1328
NM_001326326.1
NP_001313255.1
12
1253
NM_001326327.1
NP_001313256.1
12
1271
NM_001326328.1
NP_001313257.1
13
1365
NM_001326329.1
NP_001313258.1
11
1287
NM_001326330.1
NP_001313259.1
11
1305
NM_001326331.1
NP_001313260.1
11
1457
NM_001326332.1
NP_001313261.1
11
1439
NM_001326333.1
NP_001313262.1
10
1377
NM_001326334.1
NP_001313263.1
12
1642
NM_001326335.1
NP_001313264.1
11
1451
NM_001326336.1
NP_001313265.1
12
1511
NM_001326338.1
NP_001313267.1
10
1345
NM_001326339.1
NP_001313268.1
10
1363
NM_001326340.1
NP_001313269.1
12
1405
NM_001326341.1
NP_001313270.1
11
1351
NM_001326342.1
NP_001313271.1
11
1369
NM_001326343.1
NP_001313272.1
12
1423
NM_001326344.1
NP_001313273.1
11
1141
NM_001326345.1
NP_001313274.1
11
1237
NM_001326346.1
NP_001313275.1
9
1384
NM_001326347.1
NP_001313276.1
11
1561
NM_001326348.1
NP_001313277.1
11
1547
NM_001326349.1
NP_001313278.1
11
1565
NM_006561.3
NP_006552.3
11
1362
XM_005252354.4
XP_005252411.1
11
1452
XM_005252357.4
XP_005252414.1
9
1053
XM_005252358.3
XP_005252415.1
9
1394
XM_006717373.2
XP_006717436.1
10
1492
XM_011519294.2
XP_011517596.1
11
1434
XM_011519297.1
XP_011517599.1
11
1557
XM_017015542.1
XP_016871031.1
10
1140
XM_017015543.1
XP_016871032.1
11
1230
XM_017015544.1
XP_016871033.1
11
1218
XM_017015545.1
XP_016871034.1
11
1212
XM_017015546.1
XP_016871035.1
11
1212
XM_017015547.1
XP_016871036.1
11
1464
XM_017015548.1
XP_016871037.1
11
1200
XM_017015549.1
XP_016871038.1
11
1200
XM_017015550.1
XP_016871039.1
11
2198
XM_017015551.1
XP_016871040.1
11
1446
XM_017015552.1
XP_016871041.1
11
1587
XM_017015553.1
XP_016871042.1
11
1575
XM_017015554.1
XP_016871043.1
11
1557
XM_017015556.1
XP_016871045.1
10
1504
XM_017015557.1
XP_016871046.1
12
1169
XM_017015558.1
XP_016871047.1
10
1156
XM_017015559.1
XP_016871048.1
10
1492
XM_017015560.1
XP_016871049.1
10
1153
XM_017015561.1
XP_016871050.1
10
1480
XM_017015562.1
XP_016871051.1
10
1474
XM_017015563.1
XP_016871052.1
10
1461
XM_017015564.1
XP_016871053.1
9
1080
XM_017015565.1
XP_016871054.1
9
1083
XM_017015566.1
XP_016871055.1
9
1068
XM_017015567.1
XP_016871056.1
10
1208
XM_017015568.1
XP_016871057.1
12
1195
XM_017015569.1
XP_016871058.1
10
1489
XM_017015570.1
XP_016871059.1
9
1065
XM_017015571.1
XP_016871060.1
10
1477
XM_017015572.1
XP_016871061.1
10
1178
GenBank mRNA
AF036956.1
-
11
1332
AF086447.1
-
1
225
AF090693.1
-
11
1230
AF090694.1
-
11
1200
AF432906.1
-
11
1262
AK293164.1
-
11
1303
AK295511.1
-
9
1003
AK297313.1
-
9
1175
AK299548.1
-
11
1192
AK299992.1
-
11
1331
AK302681.1
-
8
949
AK314087.1
-
11
1371
AK316354.1
-
11
1162
BC036391.1
-
12
1370
5260623
BM699598.1
-
5
439
U69546.1
-
11
1149
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
regulation of heart contraction
nucleotide binding
RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0