Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006562.4 | NP_006553.2 | 2 | 551 | ||
GenBank mRNA | AJ004949.1 | - | 1 | 16 | ||
BC069156.1 | - | 2 | 552 | 7217017 | ||
BC136321.1 | - | 2 | 523 | 9020308 | ||
X90828.1 | - | 2 | 544 |