Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006563.3 | NP_006554.1 | 3 | 1304 | ||
XM_011527642.2 | XP_011525944.1 | 3 | 1314 | |||
GenBank mRNA | BC033580.1 | - | 3 | 1271 | 5201847 | |
BC040000.1 | - | 3 | 1282 | |||
U65404.1 | - | 3 | 1282 |