Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286474.1 | NP_001273403.1 | 26 | 1410 | ||
NM_001286475.1 | NP_001273404.1 | 24 | 1368 | |||
NM_006781.4 | NP_006772.3 | 23 | 1416 | |||
XM_017010182.1 | XP_016865671.1 | 16 | 937 | |||
XM_017010185.1 | XP_016865674.1 | 24 | 1441 | |||
GenBank mRNA | AK225742.1 | - | 26 | 1401 | ||
AK302176.1 | - | 24 | 1328 | |||
AK310230.1 | - | 25 | 1378 | |||
BC034774.1 | - | 23 | 1383 | 5169157 | ||
U60665.1 | - | 26 | 1437 |