Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGRRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006568.2 | NP_006559.1 | 5 | 700 | ||
XM_011536362.2 | XP_011534664.1 | 5 | 589 | |||
XM_017020931.1 | XP_016876420.1 | 6 | 791 | |||
XM_017020932.1 | XP_016876421.1 | 6 | 657 | |||
GenBank mRNA | AK090727.1 | - | 1 | 1689 | ||
AK289478.1 | - | 5 | 731 | |||
AK315318.1 | - | 5 | 741 | |||
BC015063.1 | - | 5 | 666 | 4245551 | ||
BG701924.1 | - | 3 | 435 | |||
U66469.1 | - | 5 | 636 |