Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGREF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166239.1 | NP_001159711.1 | 6 | 653 | ||
NM_006569.5 | NP_006560.3 | 6 | 804 | |||
GenBank mRNA | AK290128.1 | - | 6 | 653 | ||
AK309147.1 | - | 5 | 882 | |||
BC034764.1 | - | 6 | 780 | 5182241 | ||
U66468.1 | - | 6 | 586 |