Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322243.1 | NP_001309172.1 | 8 | 1161 | ||
NM_001728.3 | NP_001719.2 | 9 | 1513 | |||
NM_198589.2 | NP_940991.1 | 8 | 1165 | |||
NM_198590.2 | NP_940992.1 | 8 | 1220 | |||
NM_198591.2 | NP_940993.1 | 7 | 1063 | |||
XM_017027173.1 | XP_016882662.1 | 9 | 1398 | |||
GenBank mRNA | AK095912.1 | - | 7 | 1581 | ||
AK296858.1 | - | 6 | 678 | |||
AK302231.1 | - | 8 | 1033 | |||
AY358113.1 | - | 9 | 1384 | |||
GU557064.1 | - | 8 | 1187 | |||
GU557065.1 | - | 7 | 1030 | |||
L10240.1 | - | 8 | 998 |