Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006593.3 | NP_006584.1 | 3 | 1252 | ||
GenBank mRNA | AK226050.1 | - | 2 | 1588 | ||
AK297438.1 | - | 2 | 420 | |||
AK299533.1 | - | 2 | 152 | |||
AK315865.1 | - | 2 | 147 | |||
AK316161.1 | - | 2 | 283 | |||
BC029289.1 | - | 3 | 1013 | |||
BC104844.1 | - | 3 | 1180 | 8143847 | ||
BC113418.1 | - | 3 | 1180 | 8322470 | ||
U49250.1 | - | 3 | 959 |