Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199420.3 | NP_955452.3 | 27 | 7502 | ||
XM_011512343.2 | XP_011510645.1 | 26 | 7352 | |||
XM_011512347.2 | XP_011510649.1 | 27 | 7490 | |||
XM_011512348.2 | XP_011510650.1 | 26 | 7359 | |||
XM_011512354.2 | XP_011510656.1 | 15 | 5248 | |||
XM_017005565.1 | XP_016861054.1 | 21 | 6354 | |||
GenBank mRNA | AB209867.1 | - | 27 | 7429 | ||
AF043628.1 | - | 12 | 3072 | |||
AF052573.1 | - | 12 | 5302 | |||
AY032677.1 | - | 28 | 7775 | |||
AY338826.2 | - | 27 | 7502 | |||
CR936627.1 | - | 25 | 9367 |