Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGEA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142434.1 | NP_001135906.1 | 8 | 1569 | ||
NM_012215.3 | NP_036347.1 | 9 | 1728 | |||
XM_017015585.1 | XP_016871074.1 | 10 | 2105 | |||
XM_017015586.1 | XP_016871075.1 | 6 | 892 | |||
GenBank mRNA | AB014579.2 | - | 9 | 1290 | ||
AF036144.2 | - | 9 | 1680 | |||
AF307332.1 | - | 9 | 1680 | |||
AK002091.1 | - | 1 | 80 | |||
BC032591.1 | - | 7 | 1292 | |||
BC039583.2 | - | 8 | 1528 | 5558183 | ||
BC047877.1 | - | 9 | 1683 |