Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006600.3 | NP_006591.1 | 9 | 1104 | ||
XM_011540529.1 | XP_011538831.1 | 9 | 1004 | |||
XM_011540530.1 | XP_011538832.1 | 9 | 1242 | |||
XM_017000094.1 | XP_016855583.1 | 10 | 1175 | |||
XM_017000095.1 | XP_016855584.1 | 10 | 1274 | |||
XM_017000096.1 | XP_016855585.1 | 10 | 1284 | |||
GenBank mRNA | AB019408.1 | - | 9 | 1052 | ||
AF086922.1 | - | 9 | 1067 | |||
AF100760.1 | - | 9 | 1067 | |||
AF125465.1 | - | 9 | 1075 | |||
AF130736.1 | - | 9 | 1018 | |||
AF241788.1 | - | 8 | 1063 | |||
AL136725.1 | - | 9 | 1080 | |||
BC002399.2 | - | 9 | 1016 | 2822167 | ||
BC003132.1 | - | 9 | 1024 | 2958251 | ||
BC006147.1 | - | 9 | 1050 | 3954220 | ||
BC007280.1 | - | 9 | 1093 | 3343588 | ||
BC015153.1 | - | 9 | 1092 | 3883328 | ||
BC021139.1 | - | 9 | 1065 | 4871007 |