Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGES3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282601.1 | NP_001269530.1 | 2 | 452 | ||
NM_001282602.1 | NP_001269531.1 | 2 | 452 | |||
NM_001282603.1 | NP_001269532.1 | 2 | 452 | |||
NM_001282604.1 | NP_001269533.1 | 3 | 501 | |||
NM_001282605.1 | NP_001269534.1 | 2 | 452 | |||
NM_006601.6 | NP_006592.3 | 2 | 452 | |||
NR_104219.1 | 2 | 789 | ||||
XM_005268576.4 | XP_005268633.1 | 2 | 2042 | |||
XM_006719199.2 | XP_006719262.1 | 2 | 801 | |||
XM_011537773.2 | XP_011536075.1 | 2 | 360 | |||
XM_011537774.2 | XP_011536076.1 | 2 | 801 | |||
XM_017018716.1 | XP_016874205.1 | 2 | 801 | |||
GenBank mRNA | AK098214.1 | - | 2 | 789 | ||
AK291945.1 | - | 2 | 206 | |||
AK293139.1 | - | 2 | 419 | |||
AK295208.1 | - | 2 | 206 | |||
AK298147.1 | - | 2 | 328 | |||
AK298160.1 | - | 2 | 266 | |||
AL603017.1 | - | 2 | 338 | |||
BC003005.1 | - | 2 | 149 | 2821965 | ||
BM541503.1 | - | 3 | 318 | |||
BP194632.1 | - | 2 | 452 | |||
BU167210.1 | - | 2 | 307 | |||
L24804.1 | - | 2 | 275 |