Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042749.2 | NP_001036214.1 | 6 | 771 | ||
NM_001042750.1 | NP_001036215.1 | 6 | 615 | |||
NM_001042751.1 | NP_001036216.1 | 6 | 615 | |||
NM_001282418.1 | NP_001269347.1 | 6 | 594 | |||
NM_006603.4 | NP_006594.3 | 5 | 550 | |||
XM_005262357.2 | XP_005262414.1 | 5 | 640 | |||
XM_005262358.2 | XP_005262415.1 | 6 | 577 | |||
XM_005262359.3 | XP_005262416.1 | 5 | 530 | |||
XM_005262360.2 | XP_005262417.1 | 5 | 459 | |||
XM_005262361.2 | XP_005262418.1 | 5 | 587 | |||
XM_006724727.1 | XP_006724790.1 | 7 | 719 | |||
XM_006724728.3 | XP_006724791.1 | 6 | 653 | |||
XM_011531253.2 | XP_011529555.1 | 5 | 512 | |||
XM_017029232.1 | XP_016884721.1 | 6 | 577 | |||
XM_017029233.1 | XP_016884722.1 | 5 | 459 | |||
XM_017029234.1 | XP_016884723.1 | 5 | 512 | |||
XM_017029235.1 | XP_016884724.1 | 6 | 658 | |||
GenBank mRNA | AK098737.1 | - | 5 | 594 | ||
AK313559.1 | - | 5 | 442 | |||
AL831939.2 | - | 5 | 519 | |||
BC001765.1 | - | 5 | 499 | 3352347 | ||
BX640970.1 | - | 6 | 626 | |||
BX641002.1 | - | 5 | 519 | |||
BX641003.1 | - | 6 | 584 | |||
CR627425.1 | - | 6 | 593 | |||
CR749433.1 | - | 6 | 584 |