Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001309434.1 | NP_001296363.1 | 3 | 201 | ||
NM_003134.5 | NP_003125.3 | 3 | 201 | |||
GenBank mRNA | AB061546.1 | - | 3 | 188 | ||
AB451391.1 | - | 3 | 115 | |||
BC035495.2 | - | 3 | 157 | 4298087 | ||
BC071716.1 | - | 3 | 137 | 4476669 | ||
BC100031.1 | - | 3 | 141 | 2847600 | ||
DA338824.1 | - | 3 | 201 | |||
U07857.1 | - | 3 | 136 | |||
X73459.1 | - | 3 | 134 |