Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006606.2 | NP_006597.2 | 5 | 464 | ||
XM_005260652.2 | XP_005260709.1 | 4 | 428 | |||
GenBank mRNA | AF237576.1 | - | 5 | 418 | ||
AL832411.1 | - | 5 | 412 | |||
BC015938.1 | - | 5 | 464 | 3922423 |