Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323041.1 | NP_001309970.1 | 9 | 1527 | ||
NM_001323042.1 | NP_001309971.1 | 9 | 1011 | |||
NM_001323043.1 | NP_001309972.1 | 9 | 1412 | |||
NM_001323044.1 | NP_001309973.1 | 9 | 1785 | |||
NM_001323045.1 | NP_001309974.1 | 10 | 1496 | |||
NM_001323046.1 | NP_001309975.1 | 6 | 1023 | |||
NM_001323047.1 | NP_001309976.1 | 9 | 1527 | |||
NM_001323048.1 | NP_001309977.1 | 9 | 1412 | |||
NM_001323049.1 | NP_001309978.1 | 8 | 1701 | |||
NM_001323050.1 | NP_001309979.1 | 10 | 1496 | |||
NM_001323051.1 | NP_001309980.1 | 9 | 1785 | |||
NM_001323052.1 | NP_001309981.1 | 9 | 1412 | |||
NM_001323053.1 | NP_001309982.1 | 9 | 1527 | |||
NM_006608.2 | NP_006599.2 | 8 | 1316 | |||
NR_136564.1 | 10 | 1611 | ||||
NR_136565.1 | 8 | 1252 | ||||
NR_136566.1 | 9 | 1527 | ||||
NR_136567.1 | 9 | 1412 | ||||
NR_136568.1 | 9 | 1336 | ||||
GenBank mRNA | AJ011863.1 | - | 8 | 895 | ||
AK126773.1 | - | 2 | 2015 | |||
AK294749.1 | - | 7 | 882 | |||
BC000842.1 | - | 2 | 131 | |||
BC002447.2 | - | 9 | 1460 | 3346990 |