Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319676.1 | NP_001306605.1 | 8 | 1135 | ||
NM_006615.2 | NP_006606.1 | 10 | 1324 | |||
NM_016452.2 | NP_057536.1 | 9 | 1246 | |||
XM_011544017.1 | XP_011542319.1 | 10 | 1308 | |||
XM_011544018.1 | XP_011542320.1 | 9 | 1230 | |||
XM_011544019.2 | XP_011542321.1 | 8 | 1230 | |||
XM_011544020.1 | XP_011542322.1 | 10 | 1054 | |||
XM_017000098.1 | XP_016855587.1 | 9 | 1224 | |||
XM_017000099.1 | XP_016855588.1 | 10 | 1054 | |||
GenBank mRNA | AB038463.1 | - | 9 | 1215 | ||
AF022799.1 | - | 10 | 1324 | |||
AK298024.1 | - | 8 | 1074 | |||
AK309039.1 | - | 10 | 1120 | |||
AK313629.1 | - | 10 | 1308 | |||
BC027993.1 | - | 8 | 1086 | 5173512 |