Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005716.3 | NP_005707.1 | 9 | 1170 | ||
NM_202468.2 | NP_974197.1 | 8 | 1114 | |||
NM_202469.2 | NP_974198.1 | 7 | 796 | |||
NM_202470.2 | NP_974199.1 | 7 | 1026 | |||
NM_202494.2 | NP_974223.1 | 6 | 708 | |||
XM_017026147.1 | XP_016881636.1 | 8 | 1061 | |||
XM_017026148.1 | XP_016881637.1 | 8 | 825 | |||
GenBank mRNA | AF028824.1 | - | 6 | 654 | ||
AF089816.1 | - | 7 | 970 | |||
AK022585.1 | - | 6 | 885 | |||
AK093820.1 | - | 3 | 1300 | |||
AK290252.1 | - | 8 | 1073 | |||
AK290948.1 | - | 7 | 762 | |||
BC000410.2 | - | 9 | 1142 | 2960992 | ||
BC004226.2 | - | 6 | 785 | |||
BC012810.2 | - | 9 | 1142 | 2960992 | ||
BC016169.1 | - | 9 | 1142 | 3530664 | ||
CR986168.1 | - | 6 | 673 |