Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206.2 | NP_001197.1 | 1 | 1561 | ||
GenBank mRNA | AB451404.1 | - | 1 | 267 | ||
AK313631.1 | - | 1 | 342 | |||
BC069431.1 | - | 1 | 340 | 7262239 | ||
BC074879.2 | - | 1 | 316 | 30915238 | ||
BC074880.2 | - | 1 | 316 | 30915472 | ||
D31716.1 | - | 1 | 1531 |