Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020024.3 | NP_064408.2 | 3 | 445 | ||
GenBank mRNA | AK047115.1 | - | 447 | |||
AK139383.1 | - | 455 | ||||
BC031418.1 | - | 3 | 445 | 3667894 | ||
BC080311.1 | - | 3 | 446 | 5713860 |