Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145158.1 | NP_001138630.1 | 15, 16 | 1982 | ||
NM_006620.3 | NP_006611.1 | 16, 17 | 2108 | |||
GenBank mRNA | AB028961.1 | - | 11, 12 | 1337 | ||
AJ420551.1 | - | 10, 11 | 1332 | |||
AJ459826.1 | - | 16, 17 | 2094 | |||
AK024258.1 | - | 16, 17 | 2029 | |||
AK293573.1 | - | 13, 14 | 1316 | |||
AK295545.1 | - | 15, 16 | 1903 | |||
AK298336.1 | - | 13, 14 | 1511 | |||
AL137664.1 | - | 4, 5 | 1420 | |||
BC001465.2 | - | 16, 17 | 2017 | 3138910 | ||
BC040849.1 | - | 16, 17 | 2012 | 5104748 | ||
BX648033.1 | - | 13, 14 | 1773 | |||
U87791.1 | - | 16, 17 | 2095 |