Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9330182L06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172706.3 | NP_766294.2 | 2 | 276 | ||
GenBank mRNA | AK034358.1 | - | 2 | 308 | ||
AK036245.1 | - | 2 | 352 | |||
AK043026.1 | - | 2 | 340 | |||
AK045969.1 | - | 2 | 310 | |||
AK048738.1 | - | 2 | 296 | |||
AK081041.1 | - | 2 | 321 | |||
AK133610.1 | - | 2 | 277 | |||
AK134808.1 | - | 2 | 337 | |||
AK161902.1 | - | 2 | 337 | |||
AK165601.1 | - | 2 | 330 | |||
BC132290.1 | - | 2 | 252 | 40130567 | ||
BC138012.1 | - | 2 | 252 | 8861031 |