Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyctk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039586.1 | NP_001034675.1 | 3 | 571 | ||
NM_174846.4 | NP_777271.3 | 2 | 707 | |||
GenBank mRNA | AF455785.1 | - | 3 | 540 | ||
AK049425.1 | - | 2 | 688 | |||
AK050182.1 | - | 3 | 561 | |||
AK052709.1 | - | 2 | 646 | |||
BC025834.1 | - | 2 | 676 | 5132437 | ||
BC025935.1 | - | 668 | 5097665 | |||
BC030732.1 | - | 2 | 389 | 5102500 | ||
BC033063.1 | - | 2 | 683 | 5100009 | ||
BC036136.1 | - | 528 | 4971642 |