Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND11 SIRNA Assays ›
Gene Symbol:
ZMYND11
Gene Name:
zinc finger MYND-type containing 11
Gene Aliases:
BRAM1, BS69, MRD30
Chromosome Location:
Chr.10: 134340 - 254637 on Build GRCh38
UniGene ID:
Hs.292265
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s21154
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZMYND11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001202464.1
NP_001189393.1
11
1129
NM_001202465.1
NP_001189394.1
10
1369
NM_001202466.1
NP_001189395.1
11
1459
NM_001202467.1
NP_001189396.1
10
1053
NM_001202468.1
NP_001189397.1
11
1215
NM_006624.5
NP_006615.2
12
1537
NM_212479.3
NP_997644.2
12
1621
XM_005252359.4
XP_005252416.1
12
1377
XM_005252361.3
XP_005252418.1
11
1298
XM_005252362.2
XP_005252419.1
11
1462
XM_006717376.2
XP_006717439.1
11
1500
XM_017015587.1
XP_016871076.1
12
1290
XM_017015588.1
XP_016871077.1
12
1526
XM_017015589.1
XP_016871078.1
12
1367
XM_017015590.1
XP_016871079.1
12
1824
XM_017015591.1
XP_016871080.1
13
1337
XM_017015592.1
XP_016871081.1
11
1205
XM_017015593.1
XP_016871082.1
11
1645
XM_017015594.1
XP_016871083.1
11
1133
GenBank mRNA
AK294469.1
-
10
1051
AK294781.1
-
11
1141
AK296427.1
-
10
1025
AK296518.1
-
7
712
AK312570.1
-
12
1274
AY732489.1
-
8
836
BC034784.1
-
12
1402
5171494
BC091489.1
-
11
1129
6186814
DQ335452.1
-
11
1196
DQ335453.1
-
10
1034
DQ335454.1
-
10
1034
DQ335455.1
-
11
1196
X86098.1
-
12
1320
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
cell cycle
cell proliferation
viral process
covalent chromatin modification
regulation of transcription elongation from RNA polymerase II promoter
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of JNK cascade
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II regulatory region sequence-specific DNA binding
chromatin binding
transcription cofactor activity
transcription corepressor activity
protein binding
zinc ion binding
methylated histone binding
histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-pfczg:80/100.66.78.229:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline