Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 3 | 283 | ||
AF034568.1 | - | 3 | 337 | |||
AK002922.1 | - | 3 | 386 | |||
AK042132.1 | - | 2 | 1696 | |||
AK079488.1 | - | 3 | 383 | |||
AK142016.1 | - | 3 | 380 | |||
AK150497.1 | - | 3 | 390 | |||
AK151463.1 | - | 3 | 404 | |||
AK160007.1 | - | 3 | 392 | |||
AK160706.1 | - | 3 | 370 | |||
AK169305.1 | - | 3 | 356 | |||
BC043657.1 | - | 3 | 355 | 5343539 |