Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178726.3 | NP_848841.2 | 2 | 1169 | ||
GenBank mRNA | AF117832.1 | - | 1 | 124 | ||
AK028275.1 | - | 2 | 211 | |||
AK032529.1 | - | 2 | 1227 | |||
AK035912.1 | - | 2 | 1172 | |||
AK045724.1 | - | 2 | 1210 | |||
AK131646.1 | - | 2 | 1156 | |||
AK142789.1 | - | 2 | 1187 | |||
AK147389.1 | - | 2 | 1170 | |||
AK147876.1 | - | 2 | 1206 | |||
AK165160.1 | - | 2 | 1202 | |||
AK190317.1 | - | 1 | 114 | |||
AK220523.1 | - | 2 | 1174 | |||
AY184801.1 | - | 2 | 757 | |||
BC096031.1 | - | 2 | 1169 | 6837066 |