Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145211.2 | NP_660212.2 | 3 | 552 | ||
GenBank mRNA | AK149613.1 | - | 3 | 513 | ||
BC013715.1 | - | 3 | 534 | 3257027 | ||
BC057878.1 | - | 3 | 511 | 5348858 | ||
M33863.1 | - | 538 | ||||
X04958.1 | - | 3 | 538 |