Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011829.1 | NP_001011829.1 | 1 | 591 |
Gene Symbol: Olfr126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146890.2 | NP_667101.1 | 1 | 591 |