Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177551.1 | NP_001171022.1 | 2 | 1778 | ||
NM_153110.4 | NP_694750.3 | 3 | 1851 | |||
GenBank mRNA | AK054544.1 | - | 2 | 1664 | ||
AY063495.1 | - | 2 | 1613 | |||
BB709545.1 | - | 1 | 60 | |||
BC064738.1 | - | 3 | 1729 | 30049322 |