Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163643.1 | NP_001157115.1 | 10 | 1694 | ||
NM_009582.4 | NP_033608.3 | 8 | 1518 | |||
GenBank mRNA | AK029882.1 | - | 8 | 1327 | ||
AK034374.1 | - | 10 | 1510 | |||
AK081623.1 | - | 3 | 186 | |||
AK166435.1 | - | 10 | 1614 | |||
BC047158.1 | - | 7 | 1312 | 6494216 | ||
BC057572.1 | - | 10 | 1518 | 6401321 | ||
U14636.1 | - | 8 | 1350 | |||
U23789.1 | - | 7 | 1308 |