Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Greb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252071.1 | NP_001239000.1 | 15 | 2446 | ||
NM_015764.4 | NP_056579.2 | 17 | 2852 | |||
GenBank mRNA | AF180470.1 | - | 16 | 2508 | ||
AK147337.1 | - | 15 | 2380 | |||
AK157808.1 | - | 15 | 2464 | |||
BC160931.1 | - | 16 | 2378 | |||
BM932811.1 | - | 2 | 259 |