Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006627.2 | NP_006618.1 | 3 | 314 | ||
GenBank mRNA | AF001176.2 | - | 3 | 285 | ||
AK314790.1 | - | 3 | 307 | |||
BC004438.1 | - | 3 | 274 | 2820501 | ||
BC006098.1 | - | 3 | 280 | 2989197 | ||
BP245167.1 | - | 3 | 294 | |||
CR536569.1 | - | 2 | 258 | |||
Y18863.1 | - | 3 | 283 |